Learn More
STEMCELL Technologies DNase I, Size: 1 g
SDP
Supplier:  STEMCELL Technologies 1000683
Encompass
Deoxyribonuclease I (DNase I) is an endonuclease consisting of a single glycosylated polypeptide chain with two disulfide bonds. DNase is often included in tissue dissociation protocols to digest DNA that has leaked into the dissociation medium as a result of cell damage. DNase I preferentially cleaves phosphodiester linkages adjacent to pyrimidine nucleotides in both single- and double-stranded DNA yielding polynucleotides with 5’-phosphate and 3’-hydroxyl groups (Bernardi et al.). DNase I has been used for the digestion of human tissues such as microglia (Klegeris McGeer) cartilage (Dunham Koch) colon (Fukushima Fiocchi) epithelium (Fukushima Fiocchi) liver (Vatakis et al.) lung (Fujino et al.) neural (Fuja et al.) and stem cells (Kusuma et al.).
Catalog No. 50-239-0259