Learn More
Apexbio Technology LLC DNase I (GMP-grade) 2500KU
SDP
Supplier:  Apexbio Technology LLC K1088R2500
Encompass
DNase I (RNase-free) is a recombinant endonuclease expressed in E coli catalyzing the hydrolysis of single- and double-stranded DNA without sequence specificity yielding oligonucleotides carrying 5 -phosphate and 3 -hydroxyl termini Its enzymatic activity strictly depends on Ca ions while Mg or Mn ions additionally modulate enzymatic preference Mg facilitates random cleavage at any site of double-stranded DNA whereas Mn induces relatively aligned double-stranded cleavage patterns Complete heat inactivation of DNase I occurs at 65 C for 10 minutes in the presence of metal-ion chelators This RNase-free nuclease is commonly employed in RNA isolation procedures in vitro transcription assays and preparation steps prior to RT-PCR to eliminate DNA contamination The product is supplied as a purified DNase I solution free of detectable RNase activity
Catalog No. 50-000-64094
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.