Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ DNASE1 (Bovine) Recombinant Protein

Catalog No. 89942239 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2000 U
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-942-239 2000 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-942-239 Supplier Abnova™ Supplier No. P5310
Add to Cart
Add to Cart

Used for Enzyme

#REF!

Specifications

Accession Number P00639
For Use With (Application) Enzyme Activity
Formulation In 5mM Calcium Acetate, 4mg/ml glycine (stabilizer), pH 5.0 and 50% glycerol
Gene ID (Entrez) 282217
Molecular Weight (g/mol) 31.3kDa
Name DNASE1 (Bovine) Recombinant Protein
Preparation Method Yeast expression system
Quantity 2000 U
Storage Requirements Store at -20°C.
Regulatory Status RUO
Common Name DNASE1
Includes 10x reaction buffer
Gene Symbol DNASE1
Assays Deoxyribonuclease I Assay; Removing genomic DNA from RNA preparations prior to RT-PCR; Degradation of DNA templates after transcription reactions; Removing unwanted DNA from samples prior to Northern blotting; Removing DNA during biopharma and bioprocessing procedures.
Biological Activity ≥2 units per ml. One Unit causes an increase in absorbance at 260nm of 0.001 per minute per ml at 25°C when acting upon highly polymerized DNA at pH 5.0.
Species Yeast
Recombinant Recombinant
Protein Tag None
Expression System Yeast expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.