Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DNASE2B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113211
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113211 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113211 Supplier Invitrogen™ Supplier No. PA5113211
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DNASE2B Polyclonal Antibody for Western Blot, ELISA

The protein encoded by this gene shares considerable sequence similarity to, and is structurally related to DNase II. The latter is a well characterized endonuclease that catalyzes DNA hydrolysis in the absence of divalent cations at acidic pH. Unlike DNase II which is ubiquitously expressed, expression of this gene product is restricted to the salivary gland and lungs. The gene has been localized to chromosome 1p22.3 adjacent (and in opposite orientation) to the uricase pseudogene. Two transcript variants encoding different isoforms have been described for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNASE2B
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.93 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene DNASE2B
Gene Accession No. Q8WZ79
Gene Alias deoxyribonuclease DLAD; deoxyribonuclease II beta; Deoxyribonuclease-2-beta; DLAD; DNA for uricase; DNase II beta; DNase II-like acid DNase; Dnase2b; DNase2-like acid DNase; DNaseII-like acid DNase; endonuclease DLAD; lysosomal DNase II; UOX
Gene Symbols DNASE2B
Host Species Rabbit
Immunogen Recombinant Human Deoxyribonuclease-2-beta protein (68-281AA).
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 58511
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.