Learn More
ADS Biotec Inc DNASep Cartridge, 4.6 mm X 50 mm
SDP
Supplier:  ADS Biotec Inc DNA993510
Encompass
The DNASep cartridge features alkylated nonporous polystyrene-divinylbenzene (PS-DVB) copolymer microspheres for high performance nucleic acid separations. These versatile HPLC cartridges can be used for DNA mutation discovery, oligonucleotide analysis and purification, and RNA analysis and purification.
Catalog No. NC0545082