Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals DNAzol™ Reagent

Catalog No. AAJ60637AE
Change view
Click to view available options
Quantity:
50 mL
100 mL
200 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60637AE 100 mL
AAJ60637AH 200 mL
AAJ60637AD 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ60637AE Supplier Thermo Scientific Chemicals Supplier No. J60637AE
Add to Cart

DNAzol∣r Reagent, is a complete and ready-to-use electrophoretic reagent to isolate genomic DNA from solid and liquid samples of animal, plant, yeast, and bacterial origin.

DNAzol™ Reagent is used as an electrophoretic reagent for the isolation of genomic DNA from solid and liquid samples of animal and plant origin

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description

  • Cholesterol (cholesterin, cholesteryl alcohol) is a lipid sterol found in vertebrate cell membranes
  • Cholesterol is a cholestanoid, a class of molecule that contains the C-27 cholestane motif
  • A precursor of many steroids and sex hormones in animals including cortisol, aldosterone, testosterone, progesterone, and estrogen
  • It plays a role in neurotransmission and is crucial for organ systems, such as immune, reproductive and endocrine systems

Applications

  • Suitable for in vitro and in vivo applications
  • Stabilized in powder form for laboratory use only
TRUSTED_SUSTAINABILITY

Specifications

Color Blue-green
For Use With (Application) Complete and ready to use reagent for the isolation of genomic DNA from solid and liquid samples of animal and plant origin
Product Type DNA Extraction Reagent
Quantity 100 mL
Form Liquid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.