Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DNM1L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595825
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595825 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595825 Supplier Invitrogen™ Supplier No. PA595825
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: MCF-7, NIH/3T3, Mouse brain, Mouse heart, Rat brain; Cellular Location: Cytoplasm, Cytoplasmic vesicle, Endomembrane system, Golgi apparatus, Membrane, Mitochondrion outer membrane, Peripheral membrane protein, Peroxisome, clathrin-coated pit, cytosol, secretory vesicle, synaptic vesicle membrane Immunogen sequence: EEMQRIIQHC SNYSTQELLR FPKLHDAIVE VVTCLLRKRL PVTNEMVHNL VAIELAYINT KHPDFADACG LMNNNIEEQR RNRLARELPS AVSRDKSSKV PSALAPASQE PSPAASAEAD GKLIQDSRRE TKNVASGGGG VGDGVQEPTT GNWRGMLKTS KAEELLAEEK SKPIPIMPAS PQKGHAVNLL DVPVPVARKL SAREQRDCEV IERLIKSYFL IVRKNIQDSV PKAVMHFLVN HVKDTLQSEL VGQLYKSSLL DDLLTESEDM AQRRKEAADM LKALQGASQI IAEIRETHLW.

A human dynamin-related protein, DRP1 contributes to mitochondrial division in mammalian cells. It plays this important role in mitochondrial fission at steady state and during apoptosis. DRP1 is required for proper cellular distribution of mitochondria, and in mutant neurons, mitochondria are largely absent from synapses, thus providing a genetic tool to assess the role of mitochondria at synapses.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNM1L
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.59 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene DNM1L
Gene Accession No. O00429, O35303, Q8K1M6
Gene Alias 6330417M19Rik; AI450666; C-terminal region; Dlp1; dnm1l; Dnm1p/Vps1p-like protein; Dnml1; Dnmlp1; Drp1; DVLP; DYMPLE; dynamin 1 like; dynamin 1-like; dynamin family member proline-rich carboxyl-terminal domain less; Dynamin-1-like protein; Dynamin-1-like protein-like protein; Dynamin-like protein; dynamin-like protein 1; dynamin-like protein 4; Dynamin-like protein IV; dynamin-related protein 1; DYNIV 11; DYNIV-11; EC 3.6.5.5; EGK_03514; EMPF; FLJ41912; HDYNIV; N-terminal region; python; similar to dynamin 1-like; VPS1; zgc:66163
Gene Symbols DNM1L
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 437-736 of human DNM1L (NP_0361922).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10059, 114114, 74006
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.