Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DNP Monoclonal Antibody (LO-DNP-30)
GREENER_CHOICE

Catalog No. 048888
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
04-888-8 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-888-8 Supplier Invitrogen™ Supplier No. 048888
Add to Cart
Edge
Add to Cart

Rat Monoclonal Antibody

DNP Monoclonal Antibody for IHC, ELISA

DNP (2,4-dinitrophenol) is a toxic dye, chemically related to trinitrophenol (picric acid). It is used in biochemical studies of oxidative processes where it uncouples oxidative phosphorylation and is also used as a metabolic stimulant. 2,4-Dinitrophenol is used in the manufacture of dyes, wood preservatives, and as a pesticide. The acute (short-term) effects of 2,4-dinitrophenol in humans through oral exposure are nausea, vomiting, sweating, dizziness, headaches, and loss of weight. Chronic (long-term) oral exposure to 2,4-dinitrophenol in humans has resulted in the formation of cataracts and skin lesions, weight loss, and has caused effects on the bone marrow, central nervous system (CNS), and cardiovascular system. [National Center for Biotechnology Information. PubChem Compound Database; CID=1493].
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNP
Applications ELISA, Immunohistochemistry
Classification Monoclonal
Clone LO-DNP-30
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene Alias C6H3N2O4; C6H4N2O5; Dinitrophenol; DNP
Host Species Rat
Immunogen Synthetic DNP.
Purification Method Purified
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgE
Target Molecule Chemical
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.