Learn More
Enzo Life Sciences Dnp-Pro-Leu-Gly-Met-Trp-Ser-Arg-OH (5mg)
SDP
Supplier:  Enzo Life Sciences ALX260120M005
Encompass_Preferred
Substrate for MMP-2 and -9. Fluorogenic substrate for MMP-2 and MMP-9. Excitation: 280nm; emission 360nm. Formula: C44H61N13O13S. MW: 1012.1. Purity: ≥95% (HPLC). Appearance: Yellow solid. Solubility: Soluble in DMSO; dilute with distilled water or buffer. Long Term Storage: -20°C. Handling: Protect from light.
Catalog No. 50-200-8332
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.