Learn More
Selleck Chemical LLC Inaxaplin-E1288-1G
SDP
Supplier:  Selleck Chemical LLC E12881G
Encompass_Preferred
Inaxaplin (VX-147) is an orally active inhibitor of apolipoprotein L1 (APOL1) function Inaxaplin binds to APOL1 and is useful in the treatment of APOL1-associated kidney disease
Catalog No. 50-313-8656
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.