Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DOC2B Rabbit anti-Human, Mouse, Non-human Primate, Rat, Polyclonal, FabGennix

Catalog No. 502206784
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-6784 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-6784 Supplier Fabgennix Inc Supplier No. DOC2B201AP
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DOC2B Polyclonal antibody specifically detects DOC2B in Human, Mouse, Non-human Primate, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot

Specifications

Antigen DOC2B
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene DOC2B
Gene Accession No. P70169, P70610, Q14184
Gene Alias doc2b; doc2-beta; DOC2BL; double C2 domain beta; double C2 protein beta; double C2, beta; double C2-like domain-containing protein beta; double C2-like domain-containing protein beta; LOW QUALITY PROTEIN: double C2-like domain-containing protein beta; double C2-like domains, beta; OTTMUSP00000006522
Host Species Rabbit
Immunogen Synthetic Peptide taken within amino acid region 360-412 on DOC2b gene.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13447, 454397, 81820, 8447
Target Species Human, Mouse, Non-human Primate, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.