Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Docetaxel, Cytotoxic Agent, 807.9 Da, >99% Purity, 50 mg

Catalog No. 502706763
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-6763 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-6763 Supplier Abcam Supplier No. AB14124850MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Potent cytotoxic agent that disrupts microtubule assembly and induces apoptosis.

  • Molecular Weight: 807.9 Da
  • Purity: >99%
  • Potency: Potent cytotoxic agent in vitro (IC50 = 2 nM)
  • Mechanism of Action: Disrupts microtubule assembly and arrests cell division at G2/M phase; induces apoptosis
  • Applications: Antitumor agent; active in vivo and in vitro
  • Form: Solid
  • Composition: 100% Docetaxel
  • Storage: Store at +4°C

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Shipping Condition Ambient
Target Molecule Microtubules
Buffer 100% Docetaxel
Content And Storage Store at 4°C.
Form Solid
Quantity 50 mg
For Use With (Application) Antitumor agent, Cytotoxicity research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.