Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abcam Docetaxel, Cytotoxic Agent, Solid, 99% Purity, 10 mg
Description
- Molecular Weight: 807.9 Da
- Purity: >99%
- Potency: Potent cytotoxic agent in vitro (IC50 = 2 nM)
- Mechanism of Action: Disrupts microtubule assembly, arrests cell division at G2/M phase, and induces apoptosis
- Applications: Antitumor agent; active in vivo and in vitro
- Form: Solid
- Constituents: 100% Docetaxel
- CAS Number: 114977-28-5
- Storage: Store at +4°C
Specifications
Specifications
| Content And Storage | Store at 4°C. |
| Form | Solid |
| Shipping Condition | Ambient |
| Target Molecule | Microtubules |
| Formulation | 100% Docetaxel |
| Quantity | 10 mg |
| For Use With (Application) | Antitumor research, Cell cycle arrest, Apoptosis induction |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?