Learn More
Medchemexpress LLC Docetaxel | 114977-28-5 | 99.9% | 807.88 | C43H53NO14 | 10 MG

Supplier: Medchemexpress LLC HYB0011R10MG
Docetaxel (standard) is an analytical reference of the chemotherapeutic agent docetaxel supplied for research and analytical applications. It acts as a microtubule depolymerization inhibitor that arrests cells at G2/M and promotes apoptosis. The product is provided in small vial sizes for laboratory workflows and is accompanied by supporting documentation (SDS, COA).
- High purity suitable for analytical applications.
- Molecular weight 807.88.
- CAS number 114977-28-5 for unambiguous identification.
- Available in multiple small vial sizes for flexibility in experiments.
- Intended for research and analytical use only.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.