Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DOCK1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583922
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583922 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583922 Supplier Invitrogen™ Supplier No. PA583922
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ILSETISPLR SQRPKSQVMN VIGSERRFSV SPSSPSSQQT PPPVTPRAKL SFSMQSSLEL NGMTGTDVAD VPPPLPLKGS VADYGNLMEN QD Highest antigen sequence indentity to the following orthologs: Rat - 87%, Mouse - 89%.

This gene encodes a loosely associated peripheral membrane protein related to the LanC family of bacterial membrane-associated proteins involved in the biosynthesis of antimicrobial peptides. This protein may play a role as a peptide-modifying enzyme component in eukaryotic cells. Previously considered a member of the G-protein-coupled receptor superfamily, this protein is now in the LanC family. Multiple alternatively spliced variants, encoding the same protein, have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DOCK1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DOCK1
Gene Accession No. Q14185
Gene Alias 180 kDa protein downstream of CRK; 9130006G06Rik; AI854900; ced5; D630004B07Rik; dedicator of cytokinesis 1; dedicator of cyto-kinesis 1; dedicator of cytokinesis protein 1; dock 180; DOCK1; DOCK180; DOwnstream of CrK; RGD1566072; RP11-82L9.1
Gene Symbols DOCK1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DOCK1. Recombinant protein control fragment (Product #RP-103032).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1793
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.