Learn More
Abcam Docosa-4Z,7Z,10Z,13Z,16Z,19Z-Hexaenoic Acid, Purity >98%, 100 mg
Description
- Molecular Weight: 328.5 g/mol
- Purity: >98%
- Biological Activity: Anti-inflammatory agent that prevents LPS-induced dendritic cell maturation, improves nitroso-redox balance, reduces VEGF-mediated angiogenic signaling, and inhibits Aβ1-42 fibrillation and toxicity in vitro
- Target: Retinoid X receptor (RXR) ligand
- CAS Number: 6217-54-5
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Retinoid X Receptor |
| Buffer | 100% Docosa-4Z,7Z,10Z,13Z,16Z,19Z-hexaenoic acid |
| Content And Storage | Store at -20°C. |
| Shipping Condition | Ambient |
| Substrate Properties | Chemical Substrate |
| Formulation | 100% Docosa-4Z,7Z,10Z,13 |
| Quantity | 100 mg |
| For Use With (Application) | Anti-inflammatory research, Retinoid X receptor signaling |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.