Learn More
BIOGEMS INTERNATIONAL INC Docosahexaenoic acid (DHA) 250mg
SDP
Supplier:  BIOGEMS INTERNATIONAL INC 6215455250MG
Encompass
Docosahexaenoic acid is an essential omega-3 fatty acid that is the primary structural component of the nervous system, skin, testicles and retina. Among its functions are to modulate the carrier-mediated transport of glycine, taurine, and choline and its deficiency is linked with neural cell death and cognitive decline. It is reported to be a ligand for the retinoid X receptor (RXR) in mouse brain tissue.
Catalog No. 50-153-0213
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.