Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Docosenylsuccinic Anhydride 90.0+%, TCI America™
SDP

Catalog No. D096725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D096725G 25 g
D09675G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D096725G Supplier TCI America Supplier No. D096725G

Chemical Identifiers

CAS 58598-42-8
Molecular Formula C26H46O3
Molecular Weight (g/mol) 406.65
MDL Number MFCD00022609
InChI Key PYGZRGQVVVNPHG-OUKQBFOZNA-N
PubChem CID 5357433
IUPAC Name 3-[(10E)-docos-10-en-1-yl]oxolane-2,5-dione
SMILES CCCCCCCCCCC\C=C\CCCCCCCCCC1CC(=O)OC1=O

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 406.65
Percent Purity ≥90.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Docosenylsuccinic Anhydride
EINECSNumber (2)-0852&(2)-1140
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.