Learn More
Cayman Chemical HydroxybutyrATKeton Bod 96
SDP
Supplier:  Cayman Chemical 70074096 WELL
Encompass_Preferred
An active metabolite of tamoxifen inhibits PKC (IC50 8 UM for the rat enzyme) decreases iron (III) chloride and ascorbate-induced increases in TBARS in rat liver microsomes (IC50 17.5 UM) induces relaxation in isolated and washed rat aortic rings (pD2 9)
Catalog No. 50-289-6868
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.