Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Doctor Solution (Sodium Plumbite), Ricca Chemical
SDP

Catalog No. R26500004F Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R26500004F 4 L Cubitainer
R2650000500 500 mL Natural Poly Bottle
2650-32 1 L Natural Poly Bottle
2650-1 4 L Natural Poly Bottle
2650-5 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. R26500004F Supplier Ricca Chemical Company Supplier No. 26501CT
Add to Cart
Add to Cart

Specifications

Chemical Name or Material Doctor Solution
Color Orange
Name Note Sodium Plumbite
CAS 7732-18-5, 1310-73-2, 1317-36-8
CAS Min % 86.0, 10.71, 1.67
CAS Max % 89.0, 11.15, 1.74
Synonym dihydrogen oxide, dihydrogen monoxide
Grade Laboratory
Packaging Cubitainer
pH >12
Quantity 4 L
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.