Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Docusate Sodium, USP, 99-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18602313 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
2.5 kg
25 kg
Packaging:
Poly Pail
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-313 2.5 kg Poly Pail
18-602-314 25 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-602-313 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. DO1052.5KG
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DO105, 577-11-7, C20H37NaO7S

Spectrum™ Chemical Docusate Sodium, USP is used to treat occasional constipation. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 577-11-7
Molecular Formula C20H37NaO7S
Molecular Weight (g/mol) 444.56
MDL Number MFCD00012455
InChI Key APSBXTVYXVQYAB-UHFFFAOYNA-M
IUPAC Name sodium 1,4-bis[(2-ethylhexyl)oxy]-1,4-dioxobutane-2-sulfonate
SMILES [Na+].CCCCC(CC)COC(=O)CC(C(=O)OCC(CC)CCCC)S([O-])(=O)=O

Specifications

CAS Max % 100%
Quantity 2.5 kg
Ignition Residue 15.5-16.5%
Packaging Poly Pail
Percent Purity 99 to 100.5%
Grade USP
Chemical Name or Material Docusate Sodium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.