accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 577-11-7

IUPAC Name: sodium 1,4-bis[(2-ethylhexyl)oxy]-1,4-dioxobutane-2-sulfonate
Synonyms: docusate diethylhexyl sodium sulfosuccinate
Molecular Weight (g/mol): 444.56
Molecular Formula: C20H37NaO7S
InChi Key: APSBXTVYXVQYAB-UHFFFAOYNA-M
SMILES: [Na+].CCCCC(CC)COC(=O)CC(C(=O)OCC(CC)CCCC)S([O-])(=O)=O
Quantity 
  • (3)
  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (6)
Percent Purity 
  • (1)
  • (2)
  • (3)
  • (5)
  • (4)
  • (2)
Physical Form 
  • (4)
  • (3)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (2)
  • (8)
  • (2)
  • (2)
  • (5)
  • (2)
  • (3)
  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (6)
  • (1)
  • (2)
  • (3)
  • (5)
  • (4)
  • (2)
  • (4)
  • (3)
  • (2)