Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dodecanedioyl dichloride | 4834-98-4 | >98.0% | 267.19 g/mol | C12H20Cl2O2 | 5 G
SDP

Supplier:  Medchemexpress LLC HYW0879645G

Encompass_Preferred

Dodecanedioyl dichloride is the acid chloride derivative of dodecanedioic acid supplied as a reagent for organic synthesis and biochemical assays. It is used as an acylation reagent and as an intermediate in the preparation of diacyl compounds, polymers, and related derivatives.

  • Acid chloride reagent for acylation reactions and intermediate synthesis.
  • Molecular weight 267.19 g/mol; formula C12H20Cl2O2.
  • Appearance: colorless to light yellow liquid.
  • Purity >98.0% (as listed by distributors).
  • Density ~1.069 g/cm3.
  • Suitable for organic synthesis and polymer/intermediate preparation.
  • Storage: pure form -20°C (up to 3 years) or 4°C (up to 2 years); in solvent, store colder.

Catalog No. 50-002-08368


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.