Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dodecanedioyl Dichloride 98.0+%, TCI America™
SDP

Catalog No. D254425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D254425G 25 g
D25445G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D254425G Supplier TCI America Supplier No. D254425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4834-98-4
Molecular Formula C12H20Cl2O2
Molecular Weight (g/mol) 267.19
MDL Number MFCD00012243
InChI Key CNXXEPWXNDFGIG-UHFFFAOYSA-N
Synonym dodecanedioyldichloride, dodecandioyldichloride, dodecanedioyl chloride, acmc-209kct, dodecanedioic acid dichloride, 1,12-dodecanedioyl dichloride, dodecanedioyl dichloride
PubChem CID 2733242
IUPAC Name dodecanedioyl dichloride
SMILES C(CCCCCC(=O)Cl)CCCCC(=O)Cl

Specifications

Color Yellow
Quantity 25 g
UN Number 3265
Formula Weight 267.19
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Dodecanedioyl Dichloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.