Learn More
Crescent Chemical Co Inc DODECENYLSUCCINIC ACID
SDP
Supplier:  Crescent Chemical Co Inc SE20755.01
Encompass_Preferred
Dodecenylsuccinic acid anhydride pract. Hardener for epoxy resins. Size - 100G Storage Conditions - +15 °C TO +30 °C Catalog Number - 20755.01 CAS 25377-73-5
Catalog No. 50-247-535
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.