Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dodecyl 4-Hydroxybenzoate 98.0+%, TCI America™
SDP

Catalog No. H035225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H035225G 25 g
H0352500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H035225G Supplier TCI America Supplier No. H035225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2664-60-0
Molecular Formula C19H30O3
Molecular Weight (g/mol) 306.446
MDL Number MFCD00016473
InChI Key BAYSQTBAJQRACX-UHFFFAOYSA-N
Synonym dodecyl p-hydroxybenzoate, dodecylparaben, n-dodecyl 4-hydroxybenzoate, p-hydroxybenzoic acid dodecyl ester, benzoic acid, 4-hydroxy-, dodecyl ester, p-oxybenzoesaureduodecylester, benzoic acid, p-hydroxy-, dodecyl ester, p-oxybenzoesaureduodecylester german, 4-hydroxybenzoic acid dodecyl ester, laurylparaben
PubChem CID 17569
IUPAC Name dodecyl 4-hydroxybenzoate
SMILES CCCCCCCCCCCCOC(=O)C1=CC=C(C=C1)O

Specifications

Melting Point 55°C
Color White
Quantity 25 g
Formula Weight 306.45
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Dodecyl 4-Hydroxybenzoate
EINECSNumber (9)-1074
RTECSNumber DH2185600
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.