Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dodecyl Methyl Sulfide 98.0+%, TCI America™
SDP

Catalog No. D376725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D376725G 25 g
D37675G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D376725G Supplier TCI America Supplier No. D376725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3698-89-3
Molecular Formula C13H28S
Molecular Weight (g/mol) 216.43
MDL Number MFCD00015073
InChI Key KJWHJDGMOQJLGF-UHFFFAOYSA-N
Synonym dodecyl methyl sulfide, 2-thiatetradecane, 1-methylsulfanyl dodecane, methylthiododecane, methyl lauryl sulfide, dodecane, 1-methylthio, methyl n-dodecyl sulfide, dodecyl methyl sulphide, sulfide, dodecyl methyl, unii-77bo1dv08u
PubChem CID 77287
IUPAC Name 1-(methylsulfanyl)dodecane
SMILES CCCCCCCCCCCCSC

Specifications

Melting Point -2°C
Color Yellow
Boiling Point 110°C
Quantity 25 g
Formula Weight 216.43
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Dodecyl Methyl Sulfide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.