Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dodecylamine Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. D1452100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1452100G 100 g
D145225G 25 g
D1452500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D1452100G Supplier TCI America Supplier No. D1452100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 929-73-7
Molecular Formula C12H28ClN
Molecular Weight (g/mol) 221.81
MDL Number MFCD00042017
InChI Key TWFQJFPTTMIETC-UHFFFAOYSA-N
Synonym dodecylamine hydrochloride, laurylamine hydrochloride, n-dodecylamine hydrochloride, dodecan-1-amine hydrochloride, 1-dodecanamine, hydrochloride, dodecylamine, hydrochloride, unii-1v1470j2mf, 1-dodecanamine, hydrochloride 1:1, armeen 12d hydrochloride, acmc-209rjd
PubChem CID 458426
IUPAC Name dodecan-1-aminium chloride
SMILES [Cl-].CCCCCCCCCCCC[NH3+]

Specifications

Color White
Quantity 100 g
Formula Weight 221.81
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Dodecylamine Hydrochloride
RTECSNumber JR6750000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.