Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dodecylbenzenesulfonic acid sodium salt, ≥80%, MP Biomedicals™

Catalog No. ICN15788991 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15788991 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15788991 Supplier MP Biomedicals, Inc Supplier No. 0215788991
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A member of the linear alkylbenzenesulfonates

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25155-30-0
Molecular Formula C18H29NaO3S
Molecular Weight (g/mol) 348.48
MDL Number MFCD00011508
InChI Key HFQQZARZPUDIFP-UHFFFAOYSA-M
Synonym Sodium Dodecylbenzenesulfonate, Abeson Nam

Specifications

Color White
Assay Percent Range ≥80%
Quantity 1 kg
Formula Weight 348.5
Physical Form Flakes
Chemical Name or Material Dodecylbenzenesulfonic acid sodium salt
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.