accessibility menu, dialog, popup

UserName

Viewing profile as user:
SOOOH3CNa

CAS RN 25155-30-0

IUPAC Name: sodium 2-dodecylbenzene-1-sulfonate
Synonyms: dodecylbenzene sulfonic acid
Molecular Weight (g/mol): 348.48
Molecular Formula: C18H29NaO3S
InChi Key: HFQQZARZPUDIFP-UHFFFAOYSA-M
SMILES: [Na+].CCCCCCCCCCCCC1=C(C=CC=C1)S([O-])(=O)=O
Quantity 
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
Physical Form 
  • (5)
  • (6)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (6)
Grade 
  • (1)
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (8)
  • (6)
  • (2)
  • (4)
  • (6)
  • (3)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (5)
  • (6)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (6)
  • (1)
  • (6)