Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dodecylbenzenesulfonic acid, sodium salt, technical

Catalog No. AC325910250
Change view
Click to view available options
Quantity:
1 kg
25 kg
Packaging:
Fiber Drum
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC325910250 25 kg Fiber Drum
AC325910010 1 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC325910250 Supplier Thermo Scientific Chemicals Supplier No. 325910250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 25155-30-0 | C18H29NaO3S | 348.48 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25155-30-0
Molecular Formula C18H29NaO3S
Molecular Weight (g/mol) 348.48
InChI Key HFQQZARZPUDIFP-UHFFFAOYSA-M
Synonym Sodium dodecylbenzenesulfonate

Specifications

Color White to Yellow
pH 6.5 to 9.5 (1% soln.)
Infrared Spectrum Authentic
Assay Percent Range 88%
Packaging Fiber Drum
Quantity 25 kg
Formula Weight 348.48
Percent Purity ≥83%
Physical Form Crystalline Powder
Chemical Name or Material Dodecylbenzenesulfonic acid, sodium salt
Hazard Category Danger

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.