Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dodecylbenzenesulfonic Acid (soft type) (mixture) 95.0+%, TCI America™
SDP

Catalog No. D098925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D098925G 25 g
D0989500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D098925G Supplier TCI America Supplier No. D098925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 68584-22-5
Molecular Formula C18H30O3S
Molecular Weight (g/mol) 326.495
MDL Number MFCD00066677
InChI Key QJRVOJKLQNSNDB-UHFFFAOYSA-N
Synonym p-3-dodecyl benzenesulphonic acid, c10-c16 alkylbenzenesulfonic acid, 3-4-sulfophenyl dodecane, benzenesulfonic acid, 4-1-ethyldecyl, benzenesulfonic acid, c10-16-alkyl derivs., dsstox_cid_7923, 4-dodecan-3-yl benzenesulfonic acid, c10-c16 alkyl benzene sulfonic acid
PubChem CID 29249
IUPAC Name 4-dodecan-3-ylbenzenesulfonic acid
SMILES CCCCCCCCCC(CC)C1=CC=C(C=C1)S(=O)(=O)O

Specifications

Quantity 25 g
UN Number 2586
Percent Purity ≥95.0% (T)
Physical Form Liquid
Chemical Name or Material Dodecylbenzenesulfonic Acid (soft type) (mixture)
EINECSNumber (3)-1884&(3)-1907
RTECSNumber DB6600000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.