Learn More
Abcam Dodecylglucopyranoside, Non-ionic Detergent, Powder, Purity >99%, 1 g
Description
- MW: 348.5 g/mol
- Purity: >99%
- Form: Powder
- Storage Buffer: Constituents: 100% Dodecyl β-D-glucopyranoside
- Application: Non-ionic detergent for solubilization of membrane-bound proteins in their native state
- Characteristics: Long chain alkyl glycopyranoside and a classical nonionic amphiphile surfactant
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Proteins (Membrane-bound) |
| Shipping Condition | Ambient |
| Form | Powder |
| Content And Storage | Store at 4°C. |
| Buffer | 100% Dodecyl β-D-glucopyranoside |
| Formulation | 100% Dodecyl β-D-glucopyranoside |
| Quantity | 1 g |
| For Use With (Application) | solubilization of membrane-bound proteins |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.