Learn More
Enzo Life Sciences Dodecylphosphate (25mg). CAS: 2627-35-2
SDP
Supplier:  Enzo Life Sciences BMLLP1050025
Encompass_Preferred
Lysophosphatidic acid (LPA) receptor ligand. A specific agonist at LPA2 (EC50=700 nM) and antagonist at LPA Alternative name: Dihydrogen monolauryl phosphate. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (15 mg/ml warm) or ethanol (2 mg/ml warm). Long Term Storage: -20°C.
Catalog No. 50-201-1159
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.