Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dodecyltrimethylammonium Bromide, Spectrum™ Chemical
SDP

Catalog No. 18620672 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
10 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-620-672 25 g
18-620-671 5 g
18-620-673 100 g
18-620-674 10 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-620-672 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. D300925GM
Add to Cart
Add to Cart

Synonym: DTAB, Dodecyl Trimethyl Ammonium Bromide, Dctab

Chemical Identifiers

CAS 1119-94-4
Molecular Formula C15H34BrN
Molecular Weight (g/mol) 308.35
InChI Key XJWSAJYUBXQQDR-UHFFFAOYSA-M
IUPAC Name dodecyltrimethylazanium bromide
SMILES [Br-].CCCCCCCCCCCC[N+](C)(C)C

Specifications

Quantity 25 g
Percent Purity 99%
Packaging Amber Bottle
Chemical Name or Material Dodecyltrimethylammonium Bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.