Learn More
Dqe Doff-it Personal Privacy Kit
Supplier: Dqe HMDOFFIT
Doff-it's large over-garment lets patients disrobe while remaining completely covered during a chemical incident. Disrobing stops more contamination from reaching a victim's skin and reduces the inhalation hazard arising from off-gassing of volatile chemicals. Excellent for field decontamination in mass casualty incidents. For the same privacy and protection from exposure to cold after soap and water showering, use the Don-it post-decon kit. Available As Doff-it Personal Privacy Kit - Adult (HMDOFFIT) Doff-it Personal Privacy Kit - Adult, Box of 20 (HMDOFFBX) Doff-it Personal Privacy Kit - Youth (HMDOFFIT-Y) Doff-it Personal Privacy Kit - Youth, Box of 20 (HMDOFFBXY) Specifications Doff-it Personal Privacy Kit - Adult, Youth Gown is made of laminated Polypropylene non-woven, fluid resistant fabric Contents Doff-it® Personal Privacy Garment (Adult: fits size XS to 3XL, Youth: fits ages 5- 12 years old)
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.