Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DOG-1 Recombinant Rabbit Monoclonal Antibody (SP31)
GREENER_CHOICE

Catalog No. PIMA514582 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA514582 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA514582 Supplier Invitrogen™ Supplier No. MA514582
  • $1,064.00
    $874.65 / Each of 1

    Save $189.35

    18% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Recombinant Monoclonal Antibody

MA5-14582 targets Dog1 in IHC (P) applications and shows reactivity with Human samples. The MA5-14582 immunogen is synthetic peptide of human Dog1.

DOG-1, the novel marker, is expressed ubiquitously in gastrointestinal stromal tumors irrespective of kit or PDGFR alpha mutation status.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DOG-1
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone SP31
Concentration 0.057 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA and 0.1% sodium azide; pH 7.2
Gene ANO1
Gene Accession No. Q5XXA6
Gene Alias ANO1; anoctamin 1; anoctamin 1, calcium activated chloride channel; anoctamin-1; Ca2+-activated Cl- channel; calcium activated chloride channel; discovered on gastrointestinal stromal tumors protein 1; DOG1; FLJ10261; oral cancer overexpressed 2; Oral cancer overexpressed protein 2; ORAOV2; TAOS2; Tmem16a; Transmembrane protein 16A; transmembrane protein 16A (eight membrane-spanning domains); tumor-amplified and overexpressed sequence 2
Gene Symbols ANO1
Host Species Rabbit
Immunogen Synthetic peptide of human Dog1 (aa 400-500).
Purification Method Protein A
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55107
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.