Learn More
Biomatik Corporation Dog Interleukin 31 (IL31) ELISA Kit
Supplier:  Biomatik Corporation EKU12522
Encompass_Preferred
Dog Interleukin 31 (IL31) ELISA Kit for detecting IL31 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Dog (Canis familiaris; Canine). Detection Range: 7.8-500pg/ml. Sensitivity: 2.9pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: C7G0W1. Gene ID: 100302725. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4608