Learn More
Kingfisher Biotech Inc Dolphin CCL22 Recombinant Protein
SDP
Supplier:  Kingfisher Biotech Inc RP1866P025
Encompass_Preferred
The Dolphin CCL22 yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Dolphin CCL22 applications are for cell culture, ELISA standard, and Western Blot Control. Dolphin CCL22 yeast-derived recombinant protein can be purchased in multiple sizes. Dolphin CCL22 Specifications: (Molecular Weight: 8.5 kDa) (Amino Acid Sequence: TEAGPYGANV EDSICCRDYI RHPLPLRLVK YYYWTSESCR RPGVVLLTVR DREICADPRL PWVKKILQRL DQ (72)) (Gene ID: 101323189). For research use only. Made in the USA
Catalog No. 50-253-2676
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.