Learn More
Kingfisher Biotech Inc Dolphin IL-17A Recombinant Protein
SDP
Supplier:  Kingfisher Biotech Inc RP2115P100
Encompass_Preferred
The Dolphin IL-17A yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Dolphin IL-17A applications are for cell culture, ELISA standard, and Western Blot Control. Dolphin IL-17A yeast-derived recombinant protein can be purchased in multiple sizes. Dolphin IL-17A Specifications: (Molecular Weight: 14.9 kDa) (Amino Acid Sequence: GIIIPQSPGC PHVEDKNFPQ YVRLNLNILN RNVNSRRPSD YYKRSTSPWT LHRNEDSERY PSVIWEAKCS HLGCVNAEGK VDHHMNSVPI QQEILVLRRE PQHCPHSFRL EKMLVAVGCT CVTPIVHHMA (130)) (Gene ID: 101322970). For research use only. Made in the USA
Catalog No. 50-253-2412
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.