Learn More
Cayman Chemical DolutegravIr M1 5mg
SDP
Supplier:  Cayman Chemical 345085 MG
Encompass_Preferred
A KCC2 inhibitor (IC50 568 nM) selectively inhibits KCC2 over NKCC1 (IC50 50 UM) but does inhibit the activity of adenosine A1 and A3 receptors as well as inhibits activity of L-type calcium channels at the benzothiazepine site and hERG potassium channels by 50 in a panel at 10 UM decreases potassium ion uptake by 70 in HEK293 cells expressing KCC2 at 1 UM
Catalog No. 50-294-9065
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.