Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dolutegravir sodium salt, Thermo Scientific™

Catalog No. AC469770010
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC469770010 1 g
AC469772500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AC469770010 Supplier Thermo Scientific Chemicals Supplier No. 469770010
Add to Cart
This item is not returnable. View return policy

Specifications

Chemical Name or Material Dolutegravir sodium salt
CAS 1051375-19-9
Quantity 1 g
Shipping Condition No special requirements
Product Line Life science
Molecular Formula C20 H18 F2 N3 Na O5
Physical Form Powder or Crystals
Color White to Light-yellow
Purity ≥98.5%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.