Description
- Italian fashion, sport focused design
- Co-injected wire core independently adjustable nose pads
- Co-injected soft wire core adjustable TPR temples
- MAX3™, MAX6™, or BOSSMAN™ Mirror coatings
- Meets or Exceeds Z87+ & Military High Velocity
- Gun Metal/Red TPR, Clear/Blue TPR, Black/Black TPR, or Yellow/Blue TPR
Certifications
CSA
Compliance
Meets ANSI Z87.1-2015, CSA Z94.3 and MIL - PRF - 31013 standards.
Specifications
Specifications
| Lens Tint | Mirror |
| For Use With (Application) | Manufacturing |
| Frame Color | Clear |
| No. per Pack | 1 Pair |
| Lens Material | Polycarbonate |
| Frame Material | Polycarbonate |