Description
- Italian fashion, sport focused design
- Co-injected wire core independently adjustable nose pads
- Co-injected soft wire core adjustable TPR temples
- Available in 1.0, 1.5, 2.0, 2.5 Diopters MAX6™ coating on Clear or Brown Lens
- Meets or Exceeds Z87+ & Military High Velocity
Specifications
Specifications
| Lens Coating | Anti-fog |
| Lens Type | Magnifying |
| Lens Tint | Clear |
| For Use With (Application) | Manufacturing |
| Frame Color | Tortoise Shell |
| Magnification Power | 2.0 |
| No. per Pack | 1 Pair |
| Lens Material | Polycarbonate |
| Frame Material | Polycarbonate |