Learn More
Selleck Chemical LLC Donepezil HCl 25mg 120011-70-3
SDP
Supplier:  Selleck Chemical LLC S246225MG
Encompass_Preferred
Donepezil is a specific and potent AChE inhibitor for bAChE and hAChE with IC50 of 8.12 nM and 11.6 nM , respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8064
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.