Learn More
AdipoGen Donepezil hydrochloride
SDP
Supplier:  AdipoGen CDXD0437M250
Encompass_Preferred
Chemical. CAS 120011-70-3. Formula C24H29NO3 . HCl. MW 379.5 . 36.5. Synthetic. Potent non-competitive reversible acetylcholinesterase inhibitor that readily crosses the blood-brain barrier to reduce the breakdown of acetylcholine. Displays >1000-fold selectivity for acetylcholinesterase AChE over butyrylcholinesterase BuChE. It is commonly used in the treatment of Alzheimers disease to improve cognition, memory and behavior.
Catalog No. 50-256-1077
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.