Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Donkey Anti-Goat IgG Antibody, R&D Systems™
GREENER_CHOICESDP

Catalog No. AF109 Shop All Novus Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF109 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF109 Supplier R&D Systems Supplier No. AF109
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Donkey Polyclonal Antibody has been used in 7 publications

IgG Polyclonal specifically detects IgG in Goat samples. It is validated for Western Blot.

Specifications

Antigen IgG
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:5000 dilution
Gene Alias Immunoglobulin G, ImmunoglobulinG
Gene Symbols IGHG1
Host Species Donkey
Immunogen Goat IgG
Purification Method Affinity Purified
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Secondary
Gene ID (Entrez) 3500
Test Specificity Detects goat IgG heavy and light chains. In direct ELISAs, approximately 5% cross-reactivity with human IgG, mouse IgG, rabbit IgG and less than 1% cross-reactivity with chicken IgY is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Goat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Product Type Antibody
Isotype IgG
Product Line R&D Systems
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.