Learn More
Donkey anti-Goat IgG (H+L) Secondary Antibody, Biotin, R&D Systems™
Shop All Novus Biologicals Products

Description
Specifications
Specifications
| Antigen | IgG (H+L) |
| Applications | Dot Blot, ELISA, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Biotin |
| Dilution | Western Blot 1:2000 - 1:10000, ELISA 1:200000 - 1:250000, Immunohistochemistry 1:1000 - 1:5000, Dot Blot |
| Host Species | Donkey |
| Immunogen | Donkey anti-Goat IgG (H+L) Secondary Antibody [Biotin] was produced by repeated immunization with goat IgG whole molecule in donkey. |
| Purification Method | Multi-step |
| Quantity | 1 mg |
| Regulatory Status | RUO |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.