Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Donkey anti-Human IgG (H+L) Secondary Antibody, DyLight 550, Novus Biologicals™
SDP

Catalog No. NB484565 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB484565 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB484565 Supplier Novus Biologicals Supplier No. NBP151847R
Add to Cart
Add to Cart

Donkey Polyclonal Antibody

IgG (H+L) Polyclonal specifically detects IgG (H+L) in Human samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen IgG (H+L)
Applications ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate DyLight 550
Formulation 50 mM Sodium Borate
Host Species Donkey
Immunogen This Donkey anti-Human IgG (H+L) Secondary Antibody was developed against human IgG-heavy and light chain.
Purification Method Immunogen affinity purified
Quantity 1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Secondary
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.