Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Donkey anti-Human IgG (H+L) Secondary Antibody, Novus Biologicals™
SDP

Catalog No. NBP260666 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP260666 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP260666 Supplier Novus Biologicals Supplier No. NBP260666
Add to Cart
Add to Cart

Donkey Polyclonal Antibody

IgG (H+L) Polyclonal specifically detects IgG (H+L) in Human samples. It is validated for ELISA, Immunohistochemistry, Western Blot.

Specifications

Antigen IgG (H+L)
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 2.4mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000 - 1:10000, ELISA 1:20000 - 1:100000, Immunohistochemistry 1:1000 - 1:5000
Formulation 0.05M Tris Chloride, 0.15M Sodium Chloride, 0.001M Magnesium Chloride, 0.0001M Zinc Chloride, 50% (v/v) Glycerol; pH 8.0 with 10 mg/ml BSA- Immunoglobulin and Protease free with 0.01% Sodium Azide
Host Species Donkey
Immunogen Human IgG whole molecule
Purification Method Affinity Purified
Quantity 2 mg
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Secondary
Test Specificity This product was prepared from monospecific antiserum by immunoaffinity chromatography using Human IgG coupled to agarose beads followed by solid phase adsorption(s) to remove any unwanted reactivities.
Target Species Human
Content And Storage Store at 4°C.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.